Learn More
Apexbio Technology LLC DL-Carnitine HCl 461-05-2 5g
SDP
Catalog No. 5000090013
Encompass_Preferred
DL-Carnitine HCl (CAS 461-05-2) is a racemic mixture comprising both D- and L- isomers of carnitine The L-isomer facilitates the mitochondrial transport of long-chain fatty acids by mediating acyl group translocation which is essential for fatty acid -oxidation This process influences cellular energy metabolism and has been associated with modulation of oxidative stress and inflammatory pathways DL-Carnitine HCl is utilized in studies investigating lipid metabolism mitochondrial function oxidative stress responses and inflammation-related mechanisms in cellular and pharmacological research contexts
Catalog No. 50-000-90013 Supplier Apexbio Technology LLC Supplier No. B19345000
DL-Carnitine HCl (CAS 461-05-2) is a racemic mixture comprising both D- and L- isomers of carnitine The L-isomer facilitates the mitochondrial transport of long-chain fatty acids by mediating acyl group translocation which is essential for fatty acid -oxidation This process influences cellular energy metabolism and has been associated with modulation of oxidative stress and inflammatory pathways DL-Carnitine HCl is utilized in studies investigating lipid metabolism mitochondrial function oxidative stress responses and inflammation-related mechanisms in cellular and pharmacological research contexts