Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Citrulline 98.0+%, TCI America™
SDP

Catalog No. C03711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C03711G Supplier TCI America Supplier No. C03711G

Chemical Identifiers

CAS 627-77-0
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.188
MDL Number MFCD00007955
InChI Key RHGKLRLOHDJJDR-UHFFFAOYSA-N
Synonym dl-citrulline, citrullin, 2-amino-5-ureidopentanoic acid, dl-2-amino-5-ureidovaleric acid, citrulina, citrulline, dl, ornithine, n5-carbamoyl-, dl, dl-ornithine, n 5-aminocarbonyl, n 5-carbamoyl-dl-ornithine, n 5-aminocarbonyl ornithine
PubChem CID 833
ChEBI CHEBI:18211
IUPAC Name 2-amino-5-(carbamoylamino)pentanoic acid
SMILES C(CC(C(=O)O)N)CNC(=O)N

Specifications

Melting Point 222°C
Color White
Quantity 1 g
Formula Weight 175.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Citrulline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.