Learn More
Medchemexpress LLC DL-Cystathionine | 535-34-2 | 97.4% | 222.26 | 1 MG
SDP
Catalog No. 5000451594
Encompass_Preferred
DL-Cystathionine is a mixture of four isomers of cystathionine and allocystathionine. It also serves as an intermediate in the biosynthesis of the amino acid cysteine.
  • It is a mixture of four isomers of cystathionine and allocystathionine.
  • It is an intermediate in the biosynthesis of the amino acid cysteine.
  • Ships at room temperature within the continental United States.
  • Store at 4°C, protected from light.
  • When in solvent, it can be stored at -80°C for six months or at -20°C for one month, protected from light.
Catalog No. 50-004-51594 Supplier Medchemexpress LLC Supplier No. HYW009749A1MG
DL-Cystathionine is a mixture of four isomers of cystathionine and allocystathionine. It also serves as an intermediate in the biosynthesis of the amino acid cysteine.
  • It is a mixture of four isomers of cystathionine and allocystathionine.
  • It is an intermediate in the biosynthesis of the amino acid cysteine.
  • Ships at room temperature within the continental United States.
  • Store at 4°C, protected from light.
  • When in solvent, it can be stored at -80°C for six months or at -20°C for one month, protected from light.