Learn More
Medchemexpress LLC DL-Cystathionine-d4 | 146764-57-0 | 99.2% | 226.29 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW009749AS1MG
Encompass_Preferred
DL-Cystathionine-d4 is the deuterium labeled DL-Cystathionine. It is a mixture of 4 isomers of cystathionine and allocystathionine. DL-Cystathionine is also an intermediate in the biosynthesis of the amino acid cysteine.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66611