Learn More
Medchemexpress LLC DL-Cystine-d4 | 108641-83-4 | 99.8% | 244.33 | 100 MG
SDP
Catalog No. 5000364956
Encompass_Preferred
DL-Cystine-d4 is a deuterium-labeled form of DL-Cystine, which is a cysteine derivative. This compound is presented as a white to off-white solid and is stable under recommended storage conditions. It is suitable for various laboratory and manufacturing applications.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Suitable for use in laboratory chemical manufacturing.
Catalog No. 50-003-64956 Supplier Medchemexpress LLC Supplier No. 100MGHYW013940S
DL-Cystine-d4 is a deuterium-labeled form of DL-Cystine, which is a cysteine derivative. This compound is presented as a white to off-white solid and is stable under recommended storage conditions. It is suitable for various laboratory and manufacturing applications.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Suitable for use in laboratory chemical manufacturing.