Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-cystine-d6 | 352431-53-9 | MFCD01317433 | 99.8% | 246.34 | C6H6D6N2O4S2 | 10 MG
SDP

Catalog No. 5000303840
Encompass_Preferred

DL-cystine-d6 is the deuterium-labeled analog of DL-cystine supplied as a stable isotope standard for laboratory research. It is intended for analytical applications such as internal standardization, tracer studies, and quantitative mass spectrometry assays.

  • Deuterium-labeled stable isotope of DL-cystine.
  • High purity (99.81%).
  • Molecular formula C6H6D6N2O4S2; molecular weight 246.34.
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Supplied in small, preweighed quantities for analytical use.
  • For research use only; not for human or clinical use.

Catalog No. 50-003-03840 Supplier Medchemexpress LLC Supplier No. HYW013940S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-cystine-d6 is the deuterium-labeled analog of DL-cystine supplied as a stable isotope standard for laboratory research. It is intended for analytical applications such as internal standardization, tracer studies, and quantitative mass spectrometry assays.

  • Deuterium-labeled stable isotope of DL-cystine.
  • High purity (99.81%).
  • Molecular formula C6H6D6N2O4S2; molecular weight 246.34.
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Supplied in small, preweighed quantities for analytical use.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.