Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Cystine Dihydrochloride, MP Biomedicals™

Catalog No. ICN10145205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10145205 5 g
ICN10145201 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10145205 Supplier MP Biomedicals, Inc Supplier No. 0210145205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 90350-38-2 | Formula: C6H14Cl2N2O4S2 | Weight: 313.21 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90350-38-2
Molecular Formula C6H14Cl2N2O4S2
Molecular Weight (g/mol) 313.21
MDL Number MFCD00084652,MFCD00064228
InChI Key HHGZUQPEIHGQST-UHFFFAOYNA-N
Synonym cystine dihydrochloride, cytosine-retha-d-riboside hydrochloride, 3,3'-disulfanediylbis 2-aminopropanoic acid dihydrochloride, cystinhydrochlorid, cystine,dihydrochloride, dl-cystine hci, acmc-209bj2, dl-cystine dihydrochloride, dl-cystine dihydrochloride crystalline, 2-amino-3-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride
PubChem CID 12297018
IUPAC Name 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid dihydrochloride
SMILES Cl.Cl.NC(CSSCC(N)C(O)=O)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 313.33
Percent Purity ≥98%
Grade Not Available
Physical Form Powder
Chemical Name or Material DL-Cystine Dihydrochloride, FTIR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.