Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dl-α-(difluoromethyl)arginine | 69955-43-7 | MFCD18252556 | 95.0% | 224.21 | C7H14F2N4O2 | 5 MG
SDP

Catalog No. 5000222945
Encompass_Preferred

DL-α-(Difluoromethyl)arginine is a small-molecule, enzyme-activated, irreversible inhibitor of arginine decarboxylases used for biochemical and microbiological research. It has documented activity blocking arginine decarboxylase in organisms such as E. coli and Pseudomonas aeruginosa and is supplied as a solid suitable for in vitro studies.

  • Irreversible arginine decarboxylase inhibition suitable for mechanistic studies.
  • Documented activity in bacterial systems such as E. coli and Pseudomonas aeruginosa.
  • Solid, white to off-white appearance simplifies handling and weighing.
  • Solubility: insoluble or slightly soluble in DMSO (< 1 mg/mL); plan appropriate solvents.
  • Purity approximately 95.0% as supplied.
  • Store sealed, away from moisture at -20°C; in solution store at -80°C for long-term stability.

Catalog No. 50-002-22945 Supplier Medchemexpress LLC Supplier No. HY1341615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-α-(Difluoromethyl)arginine is a small-molecule, enzyme-activated, irreversible inhibitor of arginine decarboxylases used for biochemical and microbiological research. It has documented activity blocking arginine decarboxylase in organisms such as E. coli and Pseudomonas aeruginosa and is supplied as a solid suitable for in vitro studies.

  • Irreversible arginine decarboxylase inhibition suitable for mechanistic studies.
  • Documented activity in bacterial systems such as E. coli and Pseudomonas aeruginosa.
  • Solid, white to off-white appearance simplifies handling and weighing.
  • Solubility: insoluble or slightly soluble in DMSO (< 1 mg/mL); plan appropriate solvents.
  • Purity approximately 95.0% as supplied.
  • Store sealed, away from moisture at -20°C; in solution store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.