Learn More
Medchemexpress LLC DL-Dipalmitoylphosphatidylcholine-d80 (DL-DPPC-d80) | 100412-79-1 | 99.0% | 814.53 | 1 MG
SDP
Catalog No. 5000363554
Encompass_Preferred
DL-Dipalmitoylphosphatidylcholine-d80 (DL-DPPC-d80) is a deuterium labeled 16:0 (Rac)-PC. This compound serves as a tracer and an internal standard in various analytical techniques. Stable heavy isotopes, including hydrogen and carbon, are incorporated into drug molecules to assist in quantitation during drug development. Deuteration is notable for its potential influence on the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Supports NMR, GC-MS, and LC-MS applications
  • Incorporates stable heavy isotopes for quantitation in drug development
  • Affects pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-63554 Supplier Medchemexpress LLC Supplier No. HY146924S1MG
DL-Dipalmitoylphosphatidylcholine-d80 (DL-DPPC-d80) is a deuterium labeled 16:0 (Rac)-PC. This compound serves as a tracer and an internal standard in various analytical techniques. Stable heavy isotopes, including hydrogen and carbon, are incorporated into drug molecules to assist in quantitation during drug development. Deuteration is notable for its potential influence on the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Supports NMR, GC-MS, and LC-MS applications
  • Incorporates stable heavy isotopes for quantitation in drug development
  • Affects pharmacokinetic and metabolic profiles of drugs