Learn More
Cayman Chemical DL-DIthIothreItol 10g
SDP
Catalog No. 502909237
Encompass_Preferred
A reducing agent; has been used to stabilize thiols in a reduced state in various applications, including protein purification and enzymatic assays
Catalog No. 50-290-9237 Supplier Cayman Chemical Supplier No. 3588510 G
A reducing agent; has been used to stabilize thiols in a reduced state in various applications, including protein purification and enzymatic assays