Learn More
Cayman Chemical DL-DIthIothreItol 1g
SDP
Catalog No. 502911091
Encompass_Preferred
A reducing agent; has been used to stabilize thiols in a reduced state in various applications, including protein purification and enzymatic assays
Catalog No. 50-291-1091 Supplier Cayman Chemical Supplier No. 358851 G
A reducing agent; has been used to stabilize thiols in a reduced state in various applications, including protein purification and enzymatic assays