Learn More
Apexbio Technology LLC DL-Dithiothreitol,25g CAS# 3483-12-3
SDP
Supplier:  Apexbio Technology LLC B729425000
Encompass_Preferred
DL-Dithiothreitol is capable of maintaining monothiols completely in the reduced state and of reducing disulfides quantitatively. Thiol groups such as those of coenzyme A are readily oxidized in air to disulfides. To maintain these groups in the reduced state, another thiol is often added so that interchange takes place. Since DL-dithiothreitol is a highly alcohol- and water-soluble solid with little odor and little tendency to be oxidized directly by air, it proves much superior to the thiols currently used as protective reagents for sulfhydryl groups. DL-Dithiothreitol is commonly used in a variety of experiments that involve peptides or proteins, protecting sulfhydryl groups from oxidation and reducing disulfide bonds between cysteines. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0106