Learn More
Cayman Chemical DL-Dithiothreitol, Purity: ≥95%, CAS: 3483-12-3, 25 g
SDP
Catalog No. 502910615
Encompass_Preferred
A reducing agent; has been used to stabilize thiols in a reduced state in various applications, including protein purification and enzymatic assays
Catalog No. 50-291-0615 Supplier Cayman Chemical Supplier No. 3588525 G
A reducing agent; has been used to stabilize thiols in a reduced state in various applications, including protein purification and enzymatic assays