Learn More
Sigma Aldrich Fine Chemicals Biosciences DL-Dithiothreitol, 3483-12-3, MFCD00004877, 1 G
Catalog No. NC1377326
Encompass
Linear Formula: HSCH2CH(OH)CH(OH)CH2SH, Molecular Weight: 154.25, ≥98% (HPLC), ≥99.0% (titration), Synonym: threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT. An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides.
Catalog No. NC1377326 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D06321G
Linear Formula: HSCH2CH(OH)CH(OH)CH2SH, Molecular Weight: 154.25, ≥98% (HPLC), ≥99.0% (titration), Synonym: threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT. An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides.