Learn More
Cayman Chemical DL-DIthIothreItol 5g
SDP
Catalog No. 502912976
Encompass_Preferred
A reducing agent; has been used to stabilize thiols in a reduced state in various applications, including protein purification and enzymatic assays
Catalog No. 50-291-2976 Supplier Cayman Chemical Supplier No. 358855 G
A reducing agent; has been used to stabilize thiols in a reduced state in various applications, including protein purification and enzymatic assays