Learn More
Sigma Aldrich Fine Chemicals Biosciences DL-Dithiothreitol, 3483-12-3, MFCD00004877, 5 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 438195G
Encompass
Linear Formula: HSCH2CH(OH)CH(OH)CH2SH, Molecular Weight: 154.25, ≥99.0% (RT), mp: 41-44 °C, Synonym: threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT.
Catalog No. NC1631828