Learn More
Medchemexpress LLC Dl-dithiothreitol-d1 | 3483-12-3 | MFCD00064305 | 154.3 g/mol | C4H10O2S2 | 10 MG
SDP
Catalog No. 5000288170
Encompass_Preferred
DL-dithiothreitol-d1 is the deuterated form of DL-dithiothreitol (DTT), a thiol reducing agent used to reduce disulfide bonds, protect thiol groups, and denature proteins during biochemical sample preparation. This listing corresponds to the 10 MG isotopically labeled reagent provided by the supplier.
Catalog No. 50-002-88170 Supplier Medchemexpress LLC Supplier No. HY15917S10MG
DL-dithiothreitol-d1 is the deuterated form of DL-dithiothreitol (DTT), a thiol reducing agent used to reduce disulfide bonds, protect thiol groups, and denature proteins during biochemical sample preparation. This listing corresponds to the 10 MG isotopically labeled reagent provided by the supplier.