Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dl-dithiothreitol-d6 | 850153-85-4 | MFCD00042404 | 99.0% | 160.29 g·mol-1 | C4H4D6O2S2 | 10 MG
SDP

Catalog No. 5000288643
Encompass_Preferred

DL-dithiothreitol-d6 (DTT-d6) is the deuterated analogue of DL-dithiothreitol, used as a reducing agent to cleave disulfide bonds and as an isotopic internal standard in analytical and mass spectrometry applications.

  • Deuterated (d6) analogue suitable for mass spectrometry and isotopic labeling applications.
  • High chemical purity: 99.0%.
  • Molecular formula C4H4D6O2S2 and molecular weight 160.29 g·mol-1.
  • Used to reduce disulfide bonds in protein and sample preparation workflows.
  • Available in 10 mg pack size for analytical use.

Catalog No. 50-002-88643 Supplier Medchemexpress LLC Supplier No. HY15917S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-dithiothreitol-d6 (DTT-d6) is the deuterated analogue of DL-dithiothreitol, used as a reducing agent to cleave disulfide bonds and as an isotopic internal standard in analytical and mass spectrometry applications.

  • Deuterated (d6) analogue suitable for mass spectrometry and isotopic labeling applications.
  • High chemical purity: 99.0%.
  • Molecular formula C4H4D6O2S2 and molecular weight 160.29 g·mol-1.
  • Used to reduce disulfide bonds in protein and sample preparation workflows.
  • Available in 10 mg pack size for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.