Learn More
Bioworld DTT 100mM 3483-12-3, 100ml
Supplier:  Bioworld 404200011
Encompass_Preferred
100mM DTT solution in water. Molecular Biology Grade. DNAse, RNAse free. IUPAC_Name: (2S,3S)-1,4-disulfanylbutane-2,3-diol. PH Value: 3.0-5.0. RTECS Number: EK1610000. Appearance Form : Liquid, Melting Point : 38-43 °C, Boiling Point : 364.5 °C, Density : 1.302g/cm3. 100ml
Catalog No. 50-255-872