Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dl-Dithiothreitol, MP Biomedicals™

Catalog No. MP21948212 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21948212 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21948212 Supplier MP Biomedicals, Inc Supplier No. 02194821.2
Add to Cart
Add to Cart

Used to stabilize sulfhydryl-containing enzymes. Dl-Dithiothreitol, MP Biomedicals reduces protein disulfide bonds in sample buffers prior to SDS-PAGE. DTT is also used to reduce the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to brea

DTT is a Clelands reagent that acts as a protective agent for sulfhydryl groups (-SH). It quantitatively reduces disulfides (-S-S- to -SH). In this reaction the DTT is oxidized to the cyclic disulfide, which ensures the reduction of other disulfides in solution. Disulfide reduction occurs quickly at pH 8.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27565-41-9
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877,MFCD00064305
InChI Key VHJLVAABSRFDPM-UHFFFAOYNA-N
Synonym 1,4-Dithiothreitol, DTT, 1,4-Dithio-DL-threitol, Cleland Reagent racemic, (±)-threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent
IUPAC Name 1,4-disulfanylbutane-2,3-diol
SMILES OC(CS)C(O)CS

Specifications

Melting Point 40°C to 50°C
pH 4.0 to 6.0 (0.1M in H2O, 25°C) (literature)
Quantity 250 mg
Solubility Information Soluble in water (50mg/mL, clear and colorless solution), ethanol, acetone, ethylate, chloroform and ether.
Formula Weight 154.2
Grade Molecular Biology Reagent
Physical Form Powder
Recommended Storage Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.