Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Dithiothreitol, MP Biomedicals™

Catalog No. MP04856127 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04856127 100 g
MP04856126 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP04856127 Supplier MP Biomedicals, Inc Supplier No. 04856127
Add to Cart
Add to Cart

DL-Dithiothreitol, Electrophoresis grade, ≥99.5%, White Powder

  • Dithiothreitol is useful for stabilizing sulfhydryl containing enzymes.
  • Effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE.
  • DTT can also be used for reducing the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel.
  • DTT is less pungent and is less toxic than 2-mercaptoethanol.
  • DL-Dithiothreitol is also known as Clelands reagent;
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym 1,4-Dithiothreitol, DTT, 1,4-Dithio-DL-threitol, Cleland Reagent
IUPAC Name (2R,3R)-1,4-disulfanylbutane-2,3-diol
SMILES O[C@@H](CS)[C@@H](O)CS

Specifications

Melting Point 40°C to 50°C
Color White
pH 4.0 to 6.0 (0.1 M in H2O, 25°C)
Flash Point 113°C / 235.4°F (lit.)
Quantity 100 g
UN Number UN3335
Beilstein 1719757
Solubility Information Soluble in water (50mg/mL - clear and colorless solution); soluble in ethanol, acetone, ethylate, chloroform and ether.
Grade ≥99.5%,
Physical Form Powder
RTECSNumber EK1610000
storageNote1 +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.