Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Dithiothreitol Solution 1M in H2O, MilliporeSigma™ Supelco™

Catalog No. 111013992 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3992 10 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3992 Supplier MilliporeSigma Supelco Supplier No. 64656310X.5ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 12-3-3483
Density 1.035 g/mL (at 25°C)
Concentration 1 M
Linear Formula C4H10O2S2
MDL Number MFCD00004877
Synonym (+/-)-threo-1,4-Dimercapto-2,3-butanediol solution; DTT
Type Organic Standard
Quantity 10 x 0.5 mL
Formula Weight 154.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.