Learn More
Medchemexpress LLC DL-Dopa | 63-84-3 | 98.6% | 197.19 | 100 MG
SDP
Catalog No. 5000437066
Encompass_Preferred
DL-Dopa is a beta-hydroxylated derivative of phenylalanine. This product is for research use only and not sold to patients.
  • Human endogenous metabolite
  • Inhibitor
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-37066 Supplier Medchemexpress LLC Supplier No. HY113404100MG
DL-Dopa is a beta-hydroxylated derivative of phenylalanine. This product is for research use only and not sold to patients.
  • Human endogenous metabolite
  • Inhibitor
  • Solid appearance
  • White to off-white color