Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Epinephrine Crystalline MP Biomedicals

Catalog No. ICN15106480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15106480 100 g
ICN15106405 5 g
ICN15106401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15106480 Supplier MP Biomedicals, Inc Supplier No. 0215106480
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 329-65-7
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
InChI Key UCTWMZQNUQWSLP-UHFFFAOYSA-N
Synonym dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol
PubChem CID 838
ChEBI CHEBI:33568
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Quantity 100 g
Formula Weight 183.2
Physical Form Crystalline
Chemical Name or Material DL-Epinephrine
storageNote1 Store under nitrogen. Protect from light.
Recommended Storage Store at 2° to 8°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.