Learn More
Bellco Glass, Inc. Delong Frrnbach Flask w/6 Baffles, 2.8L
SDP
Catalog No. 501945564
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Features a 38mm DeLong style neck and can be used with a 38mm stainless steel closure (sold separately). MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2800mlOD x OAH: 210 x250mmCase Quantity: 3
Catalog No. 50-194-5564 Supplier Bellco Glass, Inc. Supplier No. 255402833
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Features a 38mm DeLong style neck and can be used with a 38mm stainless steel closure (sold separately). MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2800mlOD x OAH: 210 x250mmCase Quantity: 3