Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Glutamic acid, 95%

Catalog No. AAH6307536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6307536 500 g
AAH6307514 25 g
AAH6307522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6307536 Supplier Thermo Scientific Chemicals Supplier No. H6307536
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 617-65-2 | C5H9NO4 | 147.13 g/mol

Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.

Solubility
Soluble in water (miscible), HCl, ether (sparingly), ethanol (sparingly), and petroleum ether (sparingly).

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-65-2
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00063113
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym dl-glutamic acid, glutamic acid, glutamic acid, dl, glutaminsaeure, glutamic acid dl-form, dl-glutamicacid, dl-glu, +--glutamic acid, 2-aminoglutaric acid, glutamate
PubChem CID 611
ChEBI CHEBI:18237
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Beilstein 1723799
Merck Index 14,4469
Solubility Information Soluble in water (miscible),HCl,ether (sparingly),ethanol (sparingly),and petroleum ether (sparingly).
Quantity 500 g
Formula Weight 147.13
Percent Purity 95%
Chemical Name or Material DL-Glutamic acid
EINECSNumber 210-522-2
RTECSNumber LZ9648000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.