Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-glutamic acid-d3 | 96927-56-9 | 98.0% | 150.15 | C5H6D3NO4 | 25 MG
SDP

Catalog No. 5000208620
Encompass_Preferred

DL-glutamic acid-d3 is the deuterium-labeled form of DL-glutamic acid provided for research use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses.

  • CAS number 96927-56-9
  • Molecular formula C5H6D3NO4
  • Molecular weight 150.15 g/mol
  • Purity 98.0%
  • Appearance white to off-white solid
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Suitable as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Catalog No. 50-002-08620 Supplier Medchemexpress LLC Supplier No. 25MGHYW041895S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

DL-glutamic acid-d3 is the deuterium-labeled form of DL-glutamic acid provided for research use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses.

  • CAS number 96927-56-9
  • Molecular formula C5H6D3NO4
  • Molecular weight 150.15 g/mol
  • Purity 98.0%
  • Appearance white to off-white solid
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Suitable as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.