Learn More
Medchemexpress LLC Dl-glutamic acid-d5 | 14341-79-8 | 99.7% | 152.16 | C5H4D5NO4 | 1 MG
SDP
Catalog No. 5000208490
Encompass_Preferred
Deuterium-labeled DL-glutamic acid (d5) provided as a research-grade stable-isotope standard for analytical applications such as mass spectrometry and metabolic tracing. The material is characterized by its molecular formula C5H4D5NO4, molecular weight 152.16, and high chemical purity.
  • Deuterium-labeled DL-glutamic acid-d5 for isotope tracing
  • High chemical purity of 99.7%
  • Available in small research pack sizes, including 1 mg
  • Supplied with data sheet, certificate of analysis, and safety data sheet
Catalog No. 50-002-08490 Supplier Medchemexpress LLC Supplier No. HYW041895S1MG
Deuterium-labeled DL-glutamic acid (d5) provided as a research-grade stable-isotope standard for analytical applications such as mass spectrometry and metabolic tracing. The material is characterized by its molecular formula C5H4D5NO4, molecular weight 152.16, and high chemical purity.
  • Deuterium-labeled DL-glutamic acid-d5 for isotope tracing
  • High chemical purity of 99.7%
  • Available in small research pack sizes, including 1 mg
  • Supplied with data sheet, certificate of analysis, and safety data sheet