Learn More
Medchemexpress LLC DL-glutamic acid-d5 | 14341-79-8 | MFCD01074021 | 99.7% | 152.16 g/mol | C5H4D5NO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW041895S10MG
Encompass_Preferred
DL-glutamic acid-d5 is a deuterium-labeled (d5) isotopic form of DL-glutamic acid supplied as a dry analytical standard for use in mass spectrometry and isotopic tracer studies. It is provided as a small pack for laboratory use.
  • deuterated internal standard for mass spectrometry
  • high isotopic and chemical purity for accurate quantitation
  • molecular formula C5H4D5NO4 and molecular weight 152.16 g/mol
  • supplied as a 10 mg pack suitable for small-scale analytical workflows
  • commonly used in metabolic tracing and analytical method development
Catalog No. 50-002-09615