Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-Glutamic acid-d5 | 14341-79-8 | MFCD01074021 | 99.7% | 152.16 g/mol | C5H4D5NO4 | 25 MG
SDP

Catalog No. 5000209027
Encompass_Preferred

DL-Glutamic acid-d5 is the deuterium-labeled analog of DL-glutamic acid, supplied as a high-purity solid for use as an internal standard and tracer in analytical and biochemical studies.

  • Deuterium-labeled DL-glutamic acid (5 deuteriums).
  • CAS number 14341-79-8.
  • Molecular formula C5H4D5NO4; molecular weight 152.16 g/mol.
  • Purity 99.7% (as listed by supplier).
  • Supplied as a 25 MG solid quantity.
  • Intended for research use only; not for human or diagnostic use.

Catalog No. 50-002-09027 Supplier Medchemexpress LLC Supplier No. 25MGHYW041895S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Glutamic acid-d5 is the deuterium-labeled analog of DL-glutamic acid, supplied as a high-purity solid for use as an internal standard and tracer in analytical and biochemical studies.

  • Deuterium-labeled DL-glutamic acid (5 deuteriums).
  • CAS number 14341-79-8.
  • Molecular formula C5H4D5NO4; molecular weight 152.16 g/mol.
  • Purity 99.7% (as listed by supplier).
  • Supplied as a 25 MG solid quantity.
  • Intended for research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.