Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-glutamic acid-d5 | 14341-79-8 | MFCD01074021 | 99.7% | 152.16 g/mol | C5H4D5NO4 | 5 MG
SDP

Catalog No. 5000209211
Encompass_Preferred

DL-glutamic acid-d5 is the deuterium-labeled analogue of DL-glutamic acid used as an isotope-labeled standard in mass spectrometry and metabolite analysis. It is supplied as a pre-weighed solid for analytical applications.

  • deuterium-labeled internal standard for mass spectrometry.
  • high purity (99.7%).
  • molecular weight 152.16 g/mol.
  • molecular formula C5H4D5NO4.
  • available as pre-weighed vials (e.g., 5 MG).

Catalog No. 50-002-09211 Supplier Medchemexpress LLC Supplier No. HYW041895S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-glutamic acid-d5 is the deuterium-labeled analogue of DL-glutamic acid used as an isotope-labeled standard in mass spectrometry and metabolite analysis. It is supplied as a pre-weighed solid for analytical applications.

  • deuterium-labeled internal standard for mass spectrometry.
  • high purity (99.7%).
  • molecular weight 152.16 g/mol.
  • molecular formula C5H4D5NO4.
  • available as pre-weighed vials (e.g., 5 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.