Learn More
Medchemexpress LLC DL-Glutamic acid (hydrate) | 19285-83-7 | 165.14 | 100 G
SDP
Catalog No. 5000453588
Encompass_Preferred
DL-Glutamic acid hydrate is a biological material or organic compound widely used in life science research. It is suitable for various laboratory applications.
  • Widely used in life science research
  • Stable as powder for 3 years at -20°C
  • Stable as powder for 2 years at 4°C
  • Stable in solvent for 6 months at -80°C
  • Stable in solvent for 1 month at -20°C
Catalog No. 50-004-53588 Supplier Medchemexpress LLC Supplier No. HYW010108100G
DL-Glutamic acid hydrate is a biological material or organic compound widely used in life science research. It is suitable for various laboratory applications.
  • Widely used in life science research
  • Stable as powder for 3 years at -20°C
  • Stable as powder for 2 years at 4°C
  • Stable in solvent for 6 months at -80°C
  • Stable in solvent for 1 month at -20°C