Learn More
Medchemexpress LLC DL-Glutamic acid (hydrate) | 19285-83-7 | 165.14 | 25 G
SDP
Catalog No. 5000367099
Encompass_Preferred
DL-Glutamic acid (hydrate) is a biological material or organic compound widely used in life science research. It is suitable for various laboratory applications.
  • Stable under recommended storage conditions
  • Powder stores at -20°C for 3 years, 4°C for 2 years
  • In solvent stores at -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-67099 Supplier Medchemexpress LLC Supplier No. 25GHYW010108
DL-Glutamic acid (hydrate) is a biological material or organic compound widely used in life science research. It is suitable for various laboratory applications.
  • Stable under recommended storage conditions
  • Powder stores at -20°C for 3 years, 4°C for 2 years
  • In solvent stores at -80°C for 6 months, -20°C for 1 month