Learn More
Medchemexpress LLC DL-Glyceraldehyde | 56-82-6 | 90.0% | 1 ML
SDP
Catalog No. 5000339642
Encompass_Preferred
DL-Glyceraldehyde is a bioactive substance involved in cellular energy metabolism and a key intermediate in sugar metabolism pathways such as glycolysis and gluconeogenesis. It is converted by enzymes into other metabolites during glycolysis to provide energy for cells, and it participates in glucose synthesis as a precursor during gluconeogenesis. It can be used in medical research to study diseases related to sugar metabolism, including diabetes and tumors.
  • Inhibits cell migration and cell viability
  • Arrests the cell cycle at the G0/G1 phase
  • Induces cell apoptosis
  • Upregulates pro-apoptotic proteins and downregulates anti-apoptotic proteins
  • Inhibits cell glycolysis
  • Shows a more significant inhibitory effect on neuroblastoma cells compared to Chinese hamster ovary K1 cells
Catalog No. 50-003-39642 Supplier Medchemexpress LLC Supplier No. HY12874810MM/1ML
DL-Glyceraldehyde is a bioactive substance involved in cellular energy metabolism and a key intermediate in sugar metabolism pathways such as glycolysis and gluconeogenesis. It is converted by enzymes into other metabolites during glycolysis to provide energy for cells, and it participates in glucose synthesis as a precursor during gluconeogenesis. It can be used in medical research to study diseases related to sugar metabolism, including diabetes and tumors.
  • Inhibits cell migration and cell viability
  • Arrests the cell cycle at the G0/G1 phase
  • Induces cell apoptosis
  • Upregulates pro-apoptotic proteins and downregulates anti-apoptotic proteins
  • Inhibits cell glycolysis
  • Shows a more significant inhibitory effect on neuroblastoma cells compared to Chinese hamster ovary K1 cells