Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-Goitrin | 13190-34-6 | 25 MG
SDP

Catalog No. 5000420358
Encompass_Preferred

DL-Goitrin, also known as (R, S)-goitrin, is a mixture of two isomeric forms, epigoitrin (R-goitrin) and (S)-goitrin. This natural ingredient is found in cruciferous vegetables and is intended for research use only.

  • Purity: 99.79%
  • Molecular weight: 129.18
  • Chemical formula: C5H7NOS
  • Appearance: Solid, white to off-white
  • Derived from plants, specifically Brassicaceae cruciferous vegetables
  • May exhibit anti-influenza virus properties

Catalog No. 50-004-20358 Supplier Medchemexpress LLC Supplier No. 25MGHYN5056
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-Goitrin, also known as (R, S)-goitrin, is a mixture of two isomeric forms, epigoitrin (R-goitrin) and (S)-goitrin. This natural ingredient is found in cruciferous vegetables and is intended for research use only.

  • Purity: 99.79%
  • Molecular weight: 129.18
  • Chemical formula: C5H7NOS
  • Appearance: Solid, white to off-white
  • Derived from plants, specifically Brassicaceae cruciferous vegetables
  • May exhibit anti-influenza virus properties

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.