Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-hexanoylcarnitine chloride | 6920-35-0 | MFCD00151472 | 98.0% | 295.80 | C13H26ClNO4 | 100 MG
SDP

Catalog No. 5000212054
Encompass_Preferred

(±)-Hexanoylcarnitine chloride is the racemic chloride salt of hexanoylcarnitine supplied as a solid reference standard for biochemical research. It is used in metabolite identification, quantitative assays, and studies of carnitine and fatty-acid metabolism.

  • Purity 98.0%.
  • White to off-white solid.
  • Molecular weight 295.80 g/mol.
  • Soluble in water (≈250 mg/mL) with ultrasonic assistance.
  • Storage: 4°C as solid; -80°C in solvent for long-term storage.
  • Available in multiple pack sizes, including 100 mg.

Catalog No. 50-002-12054 Supplier Medchemexpress LLC Supplier No. HY100978100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(±)-Hexanoylcarnitine chloride is the racemic chloride salt of hexanoylcarnitine supplied as a solid reference standard for biochemical research. It is used in metabolite identification, quantitative assays, and studies of carnitine and fatty-acid metabolism.

  • Purity 98.0%.
  • White to off-white solid.
  • Molecular weight 295.80 g/mol.
  • Soluble in water (≈250 mg/mL) with ultrasonic assistance.
  • Storage: 4°C as solid; -80°C in solvent for long-term storage.
  • Available in multiple pack sizes, including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.