Learn More
Medchemexpress LLC DL-hexanoylcarnitine chloride | 6920-35-0 | MFCD00151472 | 98.0% | 295.80 g/mol | C13H26ClNO4 | 5 MG
SDP
Catalog No. 5000211535
Encompass_Preferred
(±)-Hexanoylcarnitine chloride is the racemic chloride salt of hexanoylcarnitine, an acylcarnitine fatty-acid metabolite used as an analytical standard and research reagent in metabolomics and toxicology studies. It is provided as a solid reagent with reported high purity for quantitative and qualitative workflows.
  • High purity (98.0%) suitable for analytical use.
  • Supplied in researcher-scale pack sizes starting at 5 mg.
  • Racemic form appropriate for comparative and method development studies.
  • Solid storage recommended at 4°C; in solvent store at -80°C for long-term stability.
  • Useful as a biomarker and reference in metabolomics and toxicity investigations.
Catalog No. 50-002-11535 Supplier Medchemexpress LLC Supplier No. HY1009785MG
(±)-Hexanoylcarnitine chloride is the racemic chloride salt of hexanoylcarnitine, an acylcarnitine fatty-acid metabolite used as an analytical standard and research reagent in metabolomics and toxicology studies. It is provided as a solid reagent with reported high purity for quantitative and qualitative workflows.
  • High purity (98.0%) suitable for analytical use.
  • Supplied in researcher-scale pack sizes starting at 5 mg.
  • Racemic form appropriate for comparative and method development studies.
  • Solid storage recommended at 4°C; in solvent store at -80°C for long-term stability.
  • Useful as a biomarker and reference in metabolomics and toxicity investigations.