Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-hexanoylcarnitine chloride | 6920-35-0 | MFCD00274130 | 98.0% | 295.80 g/mol | C13H26ClNO4 | 50 MG
SDP

Catalog No. 5000211302
Encompass_Preferred

(±)-Hexanoylcarnitine chloride is a research-grade acylcarnitine supplied as a white to off-white solid. It is used as a metabolite reference and biomarker in metabolic and toxicology studies and is intended for research use only (not for human or veterinary use).

  • Racemic (DL) hexanoylcarnitine chloride in solid form.
  • White to off-white powder appearance.
  • Purity ≥98.0%.
  • Molecular weight 295.80 g/mol; CAS 6920-35-0.
  • Suitable as a reference standard and biomarker in metabolic and toxicology research.
  • Store at 4°C; in solvent store -80°C up to 6 months or -20°C up to 1 month (sealed, protect from moisture).

Catalog No. 50-002-11302 Supplier Medchemexpress LLC Supplier No. HY10097850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Hexanoylcarnitine chloride is a research-grade acylcarnitine supplied as a white to off-white solid. It is used as a metabolite reference and biomarker in metabolic and toxicology studies and is intended for research use only (not for human or veterinary use).

  • Racemic (DL) hexanoylcarnitine chloride in solid form.
  • White to off-white powder appearance.
  • Purity ≥98.0%.
  • Molecular weight 295.80 g/mol; CAS 6920-35-0.
  • Suitable as a reference standard and biomarker in metabolic and toxicology research.
  • Store at 4°C; in solvent store -80°C up to 6 months or -20°C up to 1 month (sealed, protect from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.