Learn More
Medchemexpress LLC DL-Histidine-d3 | 344299-50-9 | 99.0% | 158.17 | 1 MG
SDP
Catalog No. 5000451684
Encompass_Preferred
DL-Histidine-d3 is a deuterium labeled DL-Histidine. Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Intended for research use only.
Catalog No. 50-004-51684 Supplier Medchemexpress LLC Supplier No. HYW010209S21MG
DL-Histidine-d3 is a deuterium labeled DL-Histidine. Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Intended for research use only.