Learn More
Medchemexpress LLC DL-Histidine-d3 | 344299-50-9 | 99.0% | 158.17 | 5 MG
SDP
Catalog No. 5000366879
Encompass_Preferred
DL-Histidine-d3 is the deuterium labeled DL-Histidine. It can be used as a tracer or as an internal standard for quantitative analysis by various spectroscopic methods.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-66879 Supplier Medchemexpress LLC Supplier No. HYW010209S25MG
DL-Histidine-d3 is the deuterium labeled DL-Histidine. It can be used as a tracer or as an internal standard for quantitative analysis by various spectroscopic methods.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS