Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Histidine 98.0+%, TCI America™
SDP

Catalog No. H01481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01481G 1 g
H014825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01481G Supplier TCI America Supplier No. H01481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4998-57-6
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00005208
InChI Key HNDVDQJCIGZPNO-UHFFFAOYSA-N
Synonym H-DL-His-OH
PubChem CID 773
ChEBI CHEBI:27570
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=C(NC=N1)CC(C(=O)O)N

Specifications

Melting Point 286°C
Color White
Quantity 1 g
Formula Weight 155.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Histidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.