Learn More
Cayman Chemical DL-Homocysteine, Purity: ≥90%, CAS: 454-29-5, 1 g
SDP
Catalog No. 502992579
Encompass_Preferred
An amino acid; inhibits ACh-induced vasodilation of precontracted isolated guinea pig pulmonary artery rings and enhances phenylephrine-induced contraction of isolated guinea pig pulmonary artery rings; inhibits endothelium-dependent ACh-induced vasodilation in the vascular bed of isolated perfused rat pancreas at 200 µM; elevated plasma levels are positively correlated with occlusive arterial diseases and atherosclerosis
Catalog No. 50-299-2579 Supplier Cayman Chemical Supplier No. 302851 G
An amino acid; inhibits ACh-induced vasodilation of precontracted isolated guinea pig pulmonary artery rings and enhances phenylephrine-induced contraction of isolated guinea pig pulmonary artery rings; inhibits endothelium-dependent ACh-induced vasodilation in the vascular bed of isolated perfused rat pancreas at 200 µM; elevated plasma levels are positively correlated with occlusive arterial diseases and atherosclerosis