Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Homocysteine, ≥95% (T), MilliporeSigma™ Supelco™

Catalog No. 111014218 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4218 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4218 Supplier MilliporeSigma Supelco Supplier No. 4492525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 454-29-5
Flash Point >113°C
Physical Form Neat
Linear Formula HSCH2 CH2 CH(NH2)COOH
MDL Number MFCD00004898
Melting Point 232°C to 233°C
Synonym 2-Amino-4-mercaptobutyric acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 135.18
Percent Purity ≥95% (T)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.