Learn More
Medchemexpress LLC DL-homocysteine thiolactone hydrochloride | 6038-19-3 | MFCD00012724 | 100.0% | 153.63 g/mol | C4H8ClNOS | 1000 G
SDP
Catalog No. 5000245323
Encompass_Preferred
DL-Homocysteine thiolactone hydrochloride is a cyclic amino acid derivative used as a research chemical; it exhibits root-growth inhibitory activity and is provided for laboratory research applications. For research use only.
  • Exhibits root-growth inhibitory activity.
  • High purity (99.97%).
  • Molecular weight 153.63 g/mol and formula C4H8ClNOS.
  • Available in bulk quantities up to 1000 G and as small-scale solutions.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-45323 Supplier Medchemexpress LLC Supplier No. HY1014041000G
DL-Homocysteine thiolactone hydrochloride is a cyclic amino acid derivative used as a research chemical; it exhibits root-growth inhibitory activity and is provided for laboratory research applications. For research use only.
  • Exhibits root-growth inhibitory activity.
  • High purity (99.97%).
  • Molecular weight 153.63 g/mol and formula C4H8ClNOS.
  • Available in bulk quantities up to 1000 G and as small-scale solutions.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
  • Intended for research use only; not for human or clinical use.