Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-homocysteine thiolactone hydrochloride | 6038-19-3 | MFCD00012724 | 100.0% | 153.63 g/mol | C4H8ClNOS | 250 G
SDP

Catalog No. 5000245395
Encompass_Preferred

DL-homocysteine thiolactone hydrochloride is a cyclic amino acid derivative supplied for laboratory research. It exhibits root-growth inhibitory activity and is provided in solid and solution forms with high reported purity for use as a biochemical reagent and synthetic intermediate.

  • CAS number 6038-19-3.
  • Molecular formula C4H8ClNOS.
  • Molecular weight 153.63 g/mol.
  • Purity 99.97%.
  • Available as solids (10 g-1,000 g) and 10 mM solution in DMSO.
  • Typical use: biochemical research and synthetic intermediate.

Catalog No. 50-002-45395 Supplier Medchemexpress LLC Supplier No. HY101404250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-homocysteine thiolactone hydrochloride is a cyclic amino acid derivative supplied for laboratory research. It exhibits root-growth inhibitory activity and is provided in solid and solution forms with high reported purity for use as a biochemical reagent and synthetic intermediate.

  • CAS number 6038-19-3.
  • Molecular formula C4H8ClNOS.
  • Molecular weight 153.63 g/mol.
  • Purity 99.97%.
  • Available as solids (10 g-1,000 g) and 10 mM solution in DMSO.
  • Typical use: biochemical research and synthetic intermediate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.