Learn More
Apexbio Technology LLC DL-Homocysteinethiolactone hydrochloride 6038-19-3 1g
SDP
Catalog No. 5000090279
Encompass_Preferred
DL-Homocysteinethiolactone hydrochloride (CAS 6038-19-3) is an amino acid derivative known to inhibit root growth by modulating plant hormone biosynthesis and signaling pathways In experimental studies this molecule disrupts key processes involved in root development making it a valuable tool for elucidating mechanisms underlying plant root formation and stress responses Its application extends to agricultural and biochemical research where it is employed to investigate the regulatory networks of root system architecture and to study plant adaptation under various stress conditions
Catalog No. 50-000-90279 Supplier Apexbio Technology LLC Supplier No. C71091000
DL-Homocysteinethiolactone hydrochloride (CAS 6038-19-3) is an amino acid derivative known to inhibit root growth by modulating plant hormone biosynthesis and signaling pathways In experimental studies this molecule disrupts key processes involved in root development making it a valuable tool for elucidating mechanisms underlying plant root formation and stress responses Its application extends to agricultural and biochemical research where it is employed to investigate the regulatory networks of root system architecture and to study plant adaptation under various stress conditions