Learn More
Medchemexpress LLC HY-W009390 5g Medchemexpress, DL-Homocystine CAS:870-93-9 Purity:>98%
SDP
Catalog No. 501934525
Encompass_Preferred
Medchemexpress, HY-W009390 5g DL-Homocystine CAS:870-93-9 DL-Homocystine is the double-bonded form of homocysteine and homocysteine is recognized as an important substance in the pathogenesis and pathophysiology of schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4525 Supplier Medchemexpress LLC Supplier No. HYW0093905G
Medchemexpress, HY-W009390 5g DL-Homocystine CAS:870-93-9 DL-Homocystine is the double-bonded form of homocysteine and homocysteine is recognized as an important substance in the pathogenesis and pathophysiology of schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.