Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Homoserine lactone hydrobromide, MP Biomedicals™

Catalog No. ICN15738905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15738905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15738905 Supplier MP Biomedicals, Inc Supplier No. 0215738905
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6305-38-0
Molecular Formula C4H8BrNO2
Molecular Weight (g/mol) 182.017
InChI Key MKLNTBLOABOJFZ-UHFFFAOYSA-N
Synonym alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr
PubChem CID 73508
IUPAC Name 3-aminooxolan-2-one;hydrobromide
SMILES C1COC(=O)C1N.Br

Specifications

Color White
Quantity 5 g
Formula Weight 182.02
Physical Form Powder
Chemical Name or Material DL-Homoserine lactone hydrobromide
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.