Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DL-Indole-3-lactic acid

Catalog No. I55081GA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I55081GA 1 g
I5508A 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I55081GA Supplier Sigma Aldrich Supplier No. I55081GA
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DL-Indole-3-lactic acid

Specifications

CAS 832-97-3
Melting Point 145°C to 146°C (lit.)
Molecular Formula C11H11NO3
Linear Formula C11H11NO3
MDL Number MFCD00005642
Synonym beta-(3-Indolyl)lactic acid; 2-Hydroxy-3-(3-indolyl)propanoic acid; 3-(3-Indolyl)-2-hydroxypropanoic acid; 3-Indolyllactic acid; Indolelactic acid
Molecular Weight (g/mol) 205.21
Percent Purity 99%
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.