Learn More
Medchemexpress LLC DL-Isocitric acid (trisodium salt) (Standard) | 1637-73-6 | 91.7% | 258.07 | 50 MG
SDP
Catalog No. 5000366599
Encompass_Preferred
DL-Isocitric acid trisodium salt (Standard) is an analytical standard intended for research and analytical applications. It is an endogenous metabolite and an intermediate product in the citric acid cycle. This compound can be used as a marker for determining the composition of isocitrates in fruit products. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard
  • Intended for research and analytical applications
  • Endogenous metabolite and intermediate product in the citric acid cycle
  • Used as a marker for isocitrates in fruit products
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-003-66599 Supplier Medchemexpress LLC Supplier No. HYW009362R50MG
DL-Isocitric acid trisodium salt (Standard) is an analytical standard intended for research and analytical applications. It is an endogenous metabolite and an intermediate product in the citric acid cycle. This compound can be used as a marker for determining the composition of isocitrates in fruit products. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard
  • Intended for research and analytical applications
  • Endogenous metabolite and intermediate product in the citric acid cycle
  • Used as a marker for isocitrates in fruit products
  • Suitable for HPLC, GC, and MS experiments